ENA447956438 Enamine

2-[4-methyl-2-(morpholin-4-yl)-1,3-thiazol-5-yl]acetic acid

  • MDL number MFCD16321942
  • Cas Number : 1225769-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204691
Availability details for
Quantity
  • Availability
  • Add to Cart