ENA447956441 Enamine

1-chloro-3-(prop-2-en-1-yloxy)benzene

  • MDL number MFCD00996144
  • Cas Number : 24824-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204695

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-204695
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart