ENA447956443 Enamine

2-[4-(2,2,2-trifluoroethyl)piperazin-1-yl]-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD17150445
  • Cas Number : 1464162-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204699
Availability details for
Quantity
  • Availability
  • Add to Cart