ENA447956448 Enamine

1-(3,4-difluorophenyl)-1,4-diazepane

  • MDL number MFCD14696085
  • Cas Number : 868064-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204711
Availability details for
Quantity
  • Availability
  • Add to Cart