ENA447956451 Enamine

3-(1-benzofuran-2-yl)-2-oxopropanoic acid

  • MDL number MFCD12171613
  • Cas Number : 1156724-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204714
Availability details for
Quantity
  • Availability
  • Add to Cart