ENA447956459 Enamine

(2S)-2-hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one

  • MDL number MFCD28383837
  • Cas Number : 1807912-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204729
Availability details for
Quantity
  • Availability
  • Add to Cart