ENA447956460 Enamine

6-chloro-2,3-difluorobenzene-1-sulfonyl chloride

  • MDL number MFCD12922581
  • Cas Number : 1208078-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204730
Availability details for
Quantity
  • Availability
  • Add to Cart