ENA447956472 Enamine

2-amino-4-cyanobenzoic acid

  • MDL number MFCD20638913
  • Cas Number : 64630-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108005
Availability details for
Quantity
  • Availability
  • Add to Cart