ENA447956473 Enamine

4-(1,3-thiazol-2-yl)butan-1-ol

  • MDL number MFCD23776566
  • Cas Number : 1506479-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204773
Availability details for
Quantity
  • Availability
  • Add to Cart