ENA447956485 Enamine

[2-(1H-pyrazol-1-yl)pyridin-4-yl]methanol

  • MDL number MFCD14606194
  • Cas Number : 1248243-78-8
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170333
Availability details for
Quantity
  • Availability
  • Add to Cart