ENA447956491 Enamine

6-chloro-2,3-difluorobenzene-1-sulfonamide

  • MDL number MFCD12922580
  • Cas Number : 1208077-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204817
Availability details for
Quantity
  • Availability
  • Add to Cart