ENA447956492 Enamine

1-cyclopropyl-2-hydroxypropan-1-one

  • MDL number MFCD24700675
  • Cas Number : 25111-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204818
Availability details for
Quantity
  • Availability
  • Add to Cart