ENA447956498 Enamine

methyl 5-chloro-3-(chlorosulfonyl)-2-methoxybenzoate

  • MDL number MFCD10485914
  • Cas Number : 932854-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204825
Availability details for
Quantity
  • Availability
  • Add to Cart