ENA447956500 Enamine

methyl(2-phenylpropan-2-yl)amine

  • MDL number MFCD19160667
  • Cas Number : 66896-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204827
Availability details for
Quantity
  • Availability
  • Add to Cart