ENA447956502 Enamine

2,6-dichloro-3-methylbenzene-1-sulfonamide

  • MDL number MFCD11650645
  • Cas Number : 872277-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204830
Availability details for
Quantity
  • Availability
  • Add to Cart