ENA447956509 Enamine

2-(2,5-dimethylphenyl)-2-phenylacetaldehyde

  • MDL number MFCD22224243
  • Cas Number : 1536376-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204838
Availability details for
Quantity
  • Availability
  • Add to Cart