ENA447956518 Enamine

N-[(2,6-dichlorophenyl)methyl]cyclopropanamine hydrochloride

  • MDL number MFCD07111193
  • Cas Number : 1158762-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204852
Availability details for
Quantity
  • Availability
  • Add to Cart