ENA447956520 Enamine

N-(2-phenylethyl)hydroxylamine

  • MDL number MFCD11227211
  • Cas Number : 3217-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204854
Availability details for
Quantity
  • Availability
  • Add to Cart