ENA447956524 Enamine

5-bromo-2-methoxy-4-methylbenzoic acid

  • MDL number MFCD21234732
  • Cas Number : 90326-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204859
Availability details for
Quantity
  • Availability
  • Add to Cart