ENA447956527 Enamine

5-methyl-2-(pyridin-3-yl)-1,3-thiazol-4-amine

  • MDL number MFCD25989761
  • Cas Number : 1549002-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204863
Availability details for
Quantity
  • Availability
  • Add to Cart