ENA447956528 Enamine

2,3,5,7-tetrachloroquinoxaline

  • MDL number MFCD19373928

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204864
Availability details for
Quantity
  • Availability
  • Add to Cart