ENA447956546 Enamine

1-(2-phenylethyl)piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD07365348
  • Cas Number : 1051369-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204894
Availability details for
Quantity
  • Availability
  • Add to Cart