ENA447956563 Enamine

2-(3,4-dichlorophenyl)-2-hydroxyacetic acid

  • MDL number MFCD13194310
  • Cas Number : 56071-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175447
Availability details for
Quantity
  • Availability
  • Add to Cart