ENA447956565 Enamine

4-(4-methoxyphenyl)-3-methylbut-2-enenitrile

  • Cas Number : 81639-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204917
Availability details for
Quantity
  • Availability
  • Add to Cart