ENA447956568 Enamine

4-methylthiolan-3-one

  • MDL number MFCD08457830
  • Cas Number : 50565-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175668
Availability details for
Quantity
  • Availability
  • Add to Cart