ENA447956574 Enamine

1-[3-(naphthalen-1-yloxy)phenyl]methanamine

  • MDL number MFCD19551852
  • Cas Number : 1409211-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204944
Availability details for
Quantity
  • Availability
  • Add to Cart