ENA447956576 Enamine

1-(3-chloro-2-methylphenyl)-1,4-diazepane

  • MDL number MFCD21114365
  • Cas Number : 1368956-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204947
Availability details for
Quantity
  • Availability
  • Add to Cart