ENA447956579 Enamine

N-(4-carbamothioylpyridin-2-yl)acetamide

  • MDL number MFCD20726787
  • Cas Number : 1239503-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204950
Availability details for
Quantity
  • Availability
  • Add to Cart