ENA447956584 Enamine

5-(2-bromoethyl)-4-methyl-1,3-thiazole

  • MDL number MFCD21876637
  • Cas Number : 671-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204961
Availability details for
Quantity
  • Availability
  • Add to Cart