ENA447956589 Enamine

methyl 4-chloro-2-(pyrimidin-2-ylsulfanyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD12524511
  • Cas Number : 1184435-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204969
Availability details for
Quantity
  • Availability
  • Add to Cart