ENA447956601 Enamine

4-(2-chloroethyl)-1,3-thiazole

  • MDL number MFCD11053921
  • Cas Number : 850851-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205020
Availability details for
Quantity
  • Availability
  • Add to Cart