ENA447956603 Enamine

diamino-1,3-thiazole-5-carbonitrile

  • MDL number MFCD19206086
  • Cas Number : 2147-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205046
Availability details for
Quantity
  • Availability
  • Add to Cart