ENA447956605 Enamine

2-(pyrimidin-2-ylsulfanyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD26827881
  • Cas Number : 1566207-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205048
Availability details for
Quantity
  • Availability
  • Add to Cart