ENA447956613 Enamine

2-tert-butylpyrimidine-5-carbaldehyde

  • MDL number MFCD09863201
  • Cas Number : 104461-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205073
Availability details for
Quantity
  • Availability
  • Add to Cart