ENA447956614 Enamine

ethyl 2-fluoro-3-methylbut-2-enoate

  • MDL number MFCD27927366
  • Cas Number : 685-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205074
Availability details for
Quantity
  • Availability
  • Add to Cart