ENA447956867 Enamine

2-chloro-6-[(pyridin-4-yl)methyl]pyridine hydrochloride

  • MDL number MFCD28383887
  • Cas Number : 1803598-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205123
Availability details for
Quantity
  • Availability
  • Add to Cart