ENA447956868 Enamine

1-(3,4-dichlorophenyl)-3-methoxypropan-1-amine hydrochloride

  • MDL number MFCD28383889
  • Cas Number : 1803587-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205153
Availability details for
Quantity
  • Availability
  • Add to Cart