ENA447956870 Enamine

N,N-dimethyl-1,2,3,4-tetrahydroisoquinoline-5-sulfonamide hydrochloride

  • MDL number MFCD28383891
  • Cas Number : 1803584-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205157
Availability details for
Quantity
  • Availability
  • Add to Cart