ENA447956871 Enamine

ethyl 1-(4-chlorophenyl)-5-ethoxy-4,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD28383892
  • Cas Number : 1803605-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205161
Availability details for
Quantity
  • Availability
  • Add to Cart