ENA447956872 Enamine

5-bromo-8-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD28383893
  • Cas Number : 1803595-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205167
Availability details for
Quantity
  • Availability
  • Add to Cart