ENA447956873 Enamine

methyl 4-hydroxyisoquinoline-3-carboxylate

  • MDL number MFCD28141336
  • Cas Number : 1108146-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205176
Availability details for
Quantity
  • Availability
  • Add to Cart