ENA447956877 Enamine

7,7-dichloro-6-methyl-3-azabicyclo[4.1.0]heptane hydrochloride

  • MDL number MFCD28383897
  • Cas Number : 1803584-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205231
Availability details for
Quantity
  • Availability
  • Add to Cart