ENA447956880 Enamine

3-amino-N-(2-hydroxyethyl)-1H-pyrazole-5-carboxamide hydrochloride

  • MDL number MFCD28383901

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205331
Availability details for
Quantity
  • Availability
  • Add to Cart