ENA447956886 Enamine

N-[3-(4-fluorobenzoyl)pyridin-2-yl]-2,2-dimethylpropanamide

  • MDL number MFCD28383907
  • Cas Number : 1803592-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205392
Availability details for
Quantity
  • Availability
  • Add to Cart