ENA447956887 Enamine

1-(1H-pyrazol-4-yl)propan-1-one hydrochloride

  • MDL number MFCD28383909
  • Cas Number : 1803601-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205432
Availability details for
Quantity
  • Availability
  • Add to Cart