ENA447956888 Enamine

2-(1-methyl-4-sulfamoyl-1H-pyrrol-2-yl)-2-oxoacetic acid

  • MDL number MFCD28397406
  • Cas Number : 1803562-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205442
Availability details for
Quantity
  • Availability
  • Add to Cart