ENA447956896 Enamine

ethyl 1H-pyrazolo[1,5-a]imidazole-7-carboxylate hydrochloride

  • MDL number MFCD28383918
  • Cas Number : 1803605-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205567
Availability details for
Quantity
  • Availability
  • Add to Cart