ENA447956900 Enamine

methyl 2-amino-3-(piperidin-1-yl)propanoate dihydrochloride

  • MDL number MFCD28383923
  • Cas Number : 412932-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-179
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205632
Availability details for
Quantity
  • Availability
  • Add to Cart