ENA447956902 Enamine

methyl 3-[(5-nitro-1,3-thiazol-2-yl)sulfonyl]propanoate

  • MDL number MFCD28383926
  • Cas Number : 1803604-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205643
Availability details for
Quantity
  • Availability
  • Add to Cart