ENA447956903 Enamine

2-[(4-aminophenyl)sulfanyl]propanoic acid hydrochloride

  • MDL number MFCD28383927
  • Cas Number : 1803584-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205653
Availability details for
Quantity
  • Availability
  • Add to Cart